missing translation for 'onlineSavingsMsg'
Learn More

Mc-MMAE, MedChemExpress

Product Code. 16466788
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466788 10 mg 10mg
16439337 5 mg 5mg
16431549 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16466788 Supplier Medchem Express Supplier No. HY15741
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mc-MMAE is a protective group (maleimidocaproyl)-conjugated monomethyl auristatin E (MMAE), which is a potent tubulin inhibitor. Mc-MMAE is a drug-linker conjugate for ADC.

Mc-MMAE is a protective group (maleimidocaproyl)-conjugated monomethyl auristatin E (MMAE), which is a potent tubulin inhibitor. Mc-MMAE is a drug-linker conjugate for ADC.

Specifications

Chemical Name or Material Mc-MMAE
CAS 863971-24-8
Color White
Purity Grade Notes Research
Molecular Formula C49H78N6O10
Quantity 10 mg
Synonym Maleimidocaproyl-monomethylauristatin E
Solubility Information DMSO : 180 mg/mL (197.55 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1N(CCCCCC(N(C)[C@H](C(N[C@@H](C(C)C)C(N([C@@H]([C@@H](C)CC)[C@@H](CC(N2[C@H]([C@H](OC)[C@H](C(N[C@@H]([C@H](C3=CC=CC=C3)O)C)=O)C)CCC2)=O)OC)C)=O)=O)C(C)C)=O)C(C=C1)=O
Molecular Weight (g/mol) 911.18
Formula Weight 911.18
Percent Purity 96.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.