missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Mc-Val-Cit-PABC-PNP

Product Code. 16418018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418018 Supplier MedChemExpress Supplier No. HY20336

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mc-Val-Cit-PABC-PNP is a cathepsin cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).

Specifications

Purity 95.06%
CAS 159857-81-5
Content And Storage -20°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 737.76 g/mol
Product Type Linker
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 37 mg/mL (50.15 mM)
Synonym Maleimidocaproyl-L-valine-L-citrulline-p-aminobenzyl alcohol p-nitrophenyl carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.