missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD 100 Single-Parameter Photometer: COD

Provides accurate and reproducible results very quickly. Lovibond MD 100 Photometer uses high quality interference filters with long-life LEDs as a light source without any moving parts in a transparent sample chamber. Other features include ease of operation, ergonomic design, compact dimensions and safe handling.

Brand:  Lovibond™ 276120

Code : NEW

Additional Details : Weight : 1.00000kg

Product Code. 12671735

  • 575.00 € / Each
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual

Description

Description

The calibration and software-based adjustment options mean that the MD 100 is also suitable for use as a testing instrument. The tests are conducted using either Lovibond tablet reagents with long term stability and a guaranteed minimum five or 10 year shelf life, VARIO powder reagents or using liquid reagents.
  • Waterproof (as defined in IP68, 1 hour at 0.1m)
  • Automatic switch-off
  • Real-time clock and date calibration mode
  • Backlit LCD display (on keypress)
  • Factory calibration and user calibration; Reset to factory calibration possible
  • Internal ring memory for 16 data sets
  • Infrared interface for test data transfer
Tube Tests
These enable the user to easily perform highly sensitive and precise water testing. When using tube tests measurement is considerably faster and easier, particularly in the case of standard and serial tests. The tube tests contain a precisely measured amount of reagent, thereby avoiding the presence of superfluous chemicals and optimizing test safety. Up to six different measuring ranges are available for the various tests. The tubes are made of special optical glass with a 16mm diameter. They are supplied in a storage and dispatch box together with the digestion or auxiliary reagents. This packaging unit contains 24 or 25 reaction vials and up to two zero vials for photometer system calibration.

Specifications
  • Range: 0 to 150mg/L O2, 0 to 1,500mg/L O2, 0 to 15,000mg/L O2
  • Light source: LED, interference filter (IF) and photosensor in transparent cell chamber
  • Wavelength Accuracy: ±1nm
  • Photometric Accuracy: 3% FS (T = 20°C to 25°C)
  • Photometric Resolution: 0.01 A
  • Temperature: 5° to 40°C
  • Relative Humidity: 30 to 90% (non condensing)
  • Power Supply: 4 AAA batteries
  • Operating Time: approx. 17 hours or 5000 test measurements in continuous mode when display backlight is off
  • Dimensions (L x W x H): 15.5 x 7.5 x 3.5cm (6.1 x 3 x 1.5 in.)
  • Weight: basic unit approx. 260g
Note
Specifications for each type of tablet as well as a ‘Certificate of Analysis’ for each lot is available in the download area at www.lovibond.com.
Specifications

Specifications

MD 100 Photometer: COD (Test Tube)
3% FS
Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual
Internal ring memory for 16 data sets
0 to 150mg/L O2, 0 to 1,500mg/L O2, 0 to 15,000mg/L O2
260 g
Backlit LCD
3.5 cm
15.5 cm
LED
0.01 A
7.5 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : acide sulfurique, sulfate de mercure, su
  • REA17-CMR1-Restricted to professional users.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers