Learn More
Lovibond™ MD200 2-in-1 photometer kit
Product Code. 12651715
Change view
Click to view available options
Product Type:
Photometer
Unit Size:
Each
Product Code. Product Type Unit Size
12651715 Photometer Each
1 options
Product Code. 12651715 Supplier Lovibond™ Supplier No. 2889402
  • ONLINE EXCLUSIVE PRICE
    655.00 €
    596.82 € / Pack of 1
    Save 58.18 €
    8% Off
View all promotions
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MD200 2-in-1 photometer kit for chlorine and pH with tablet reagents includes tablet reagents 3 round vials with lids carry case batteries and instruction manual
Versatile, compact and combination of high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic results in precise measurement results at all times.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Product Type Photometer
Display Type LCD
For Use With (Application) Chlorine and pH Measurement