missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD200 3-in-1 photometer kit for chlorine pH and cyanuric acid with liquid reagents

Product Code. 12601725
Change view
Click to view available options
Product Type:
Photometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12601725 Photometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12601725 Supplier Lovibond™ Supplier No. 2882002
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

includes liquid and tablet reagents, 3 round vials with lids, carry case, batteries and instruction manual

Versatile, compact and: The benchtop photometer MD 200 combines high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic. The result: precise measurement results at all times - no matter where you use it.

Versatile use

The MD 200 photometer can be used in almost all areas of water analysis.

Precise and low maintenance

The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.

User friendly

The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.

Reagents

Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.

Specifications

Product Type Photometer
Display Type LCD
For Use With (Application) Chlorine, pH and Cyanuric Acid Measurement
compliance-icons compliance-icons compliance-icons
Product Identifier
  • MD200 photometer, range mg/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid, N,N-dimethylbenzene-1,4-diammonium sulphate,butan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.