Learn More
Lovibond™ MD200 4-in-1 photometer kit for chlorine pH cyanuirc acid and M-alkalinity with liquid reagents
Product Code. 12691725
Change view
Click to view available options
Product Type:
Photometer
Unit Size:
Each
Product Code. Product Type Unit Size
12691725 Photometer Each
1 options
Product Code. 12691725 Supplier Lovibond™ Supplier No. 2860542
  • ONLINE EXCLUSIVE PRICE
    740.00 €
    732.10 € / Pack of 1
    Save 7.90 €
    1% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
includes liquid and tablet reagents, 3 round vials with lids, carry case, batteries and instruction manual
Versatile, compact and: The benchtop photometer MD 200 combines high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic. The result: precise measurement results at all times - no matter where you use it.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Product Type Photometer
Display Type LCD
For Use With (Application) Chlorine, pH, Cyanuirc Acid and M-Alkalinity Measurement