Learn More
Lovibond™ MD200 5-in-1 photometer kit for chlorine pH cyanuric acid M-alkalinity
Product Code. 12611735
Change view
Click to view available options
Product Type:
Photometer
Unit Size:
Each
Product Code. Product Type Unit Size
12611735 Photometer Each
1 options
Product Code. 12611735 Supplier Lovibond™ Supplier No. 2861202
  • ONLINE EXCLUSIVE PRICE
    725.00 €
    716.30 € / Pack of 1
    Save 8.70 €
    1% Off
View all promotions
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
More than 400 color discs are available for the Comparator 2000+.
Versatile, compact and combination of high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic results in precise measurement results at all times.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Product Type Photometer
Display Type LCD
For Use With (Application) Chlorine, pH, Cyanuric Acid, M-Alkalinity and Calcium Hardness Measurement