missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD200 6-in-1 photometer kit for chlorine copper pH cyanuric acid M-alkalinity and iron

Product Code. 12631735
Change view
Click to view available options
Product Type:
Photometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12631735 Photometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12631735 Supplier Lovibond™ Supplier No. 2862102
  • ONLINE EXCLUSIVE PRICE
    831.00 €
    822.50 € / Pack of 1
    Save 8.50 €
    1% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MD200 6-in-1 photometer kit for chlorine copper pH cyanuric acid M-alkalinity and iron with tablet reagents 3 round vials carry case batteries and instruction manual

Versatile, compact and combination of high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic results in precise measurement results at all times.

Versatile use

The MD 200 photometer can be used in almost all areas of water analysis.

Precise and low maintenance

The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.

User friendly

The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.

Reagents

Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.

Specifications

Product Type Photometer
Display Type LCD
For Use With (Application) Chlorine, Bromine, pH, Cyanuric Acid, M-Alkalinity and Calcium Hardness Measurement
compliance-icons
Product Identifier
  • MD200 photometer, range mg / l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
Supplemental information
  • MIXTURE LIST-Contain: 2,2'- Bichinolin-4,4doacétate de calcium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.