missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Me VH 032, amine

Product Code. 18707376
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18707376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18707376 Supplier Tocris Bioscience™ Supplier No. FAB9610U100UG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Amine functionalized VHL ligand for PROTAC™ research

Me VH 032, amine is a derivative of the von Hippel-Lindau (VHL) ligand, VH 032 for use as a precursor to a PROTAC® that hijacks VHL as the E3 ubiquitin ligase component. Me VH 032, amine is supplied with a primary amine functional handle, for ready conjugation to a linker/target protein ligand. Introduction of the methyl group at the benzylic position of VH 032 is associated with improved binding affinity.

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

Require a different derivative? Get in touch

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4R)-3-Methyl-L-valyl-4-hydroxy-N-[(1S)-1-[4-(4-methyl-5-thiazolyl)phenyl]ethyl]-L-prolinamide dihydrochloride
CAS 2504950-56-3
Quantity 100 mg
Target Degrader Building Blocks
Molecular Formula C23H32N4O3S.2HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.