Learn More
WTW™ Measuring Vessel
Product Code. 10084803
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
10084803 Each
1 options
Product Code. 10084803 Supplier WTW™ Supplier No. 209023
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
TRUSTED_SUSTAINABILITY
Specifications
Type Measuring Vessel
Includes Rack and Seal, 2 x GK 600 Rubber Sleeve, 4 x DV/MG/KL Clamps and Beaker MB 50 (50mL)