missing translation for 'onlineSavingsMsg'
Learn More

Meconin, TRC

Product Code. 30010288
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010288 25 mg 25mg
17903975 5 mg 5mg
17962075 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010288 Supplier Toronto Research Chemicals Supplier No. TRCM20290025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-31-3
Molecular Formula C10 H10 O4
Molecular Weight (g/mol) 194.18
Synonym 1(3H)-Isobenzofuranone, 6,7-dimethoxy-, Phthalide, 6,7-dimethoxy- (6CI,7CI,8CI), 6,7-Dimethoxy-1(3H)-isobenzofuranone, 6,7-Dimethoxyphthalide, Meconic lactone, Meconin, Meconine, NSC 35547, Opianyl
IUPAC Name 6,7-dimethoxy-3H-2-benzofuran-1-one
SMILES COc1ccc2COC(=O)c2c1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H10O4/c1-12-7-4-3-6-5-14-10(11)8(6)9(7)13-2/h3-4H,5H2,1-2H3
Formula Weight 194.0579
Chemical Name or Material Meconin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.