missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Medetomidine hydrochloride
GREENER_CHOICE

Product Code. 18700939
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18700939 10 mg 10mg
30002072 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18700939 Supplier Tocris Bioscience™ Supplier No. 5160/10
  • ONLINE EXCLUSIVE PRICE
    224.00 €
    211.05 € / 10mg
    Save 12.95 €
    5% Off
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and highly selective α2 agonist

Potent, highly selective α2-adrenoceptor agonist (Ki values are 1.08 and 1750 nM for α2- and α1-adrenoceptors respectively). Displays greater selectivity over α1-adrenoceptors than clonidine and UK 14,304 (1620-, 220- and 300-fold respectively). Inhibits twitch response in electrically stimulated mouse vas deferens (pD2 = 9.0). Active in vivo; displays hypotensive, bradycardic, sedative, anxiolytic, hypothermic and analgesic effects. Active Isomer also available.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86347-15-1
Molecular Formula C13H17ClN2

Specifications

Quantity 10 mg
Formula Weight 236.74
Molecular Weight (g/mol) 236.743
Chemical Name or Material Medetomidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.