Learn More
Ansell MEDI-GRIP Latex Surgical Glove
Product Code. 17784949 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Unit Size:
50 pairs
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17784949 8 50 pairs
17734949 5.5 50 pairs
17744949 6 50 pairs
17754949 6.5 50 pairs
17764949 7 50 pairs
17774949 7.5 50 pairs
17794949 8.5 50 pairs
17704959 9 50 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17784949 Supplier Ansell Supplier No. 330200780
  • ONLINE EXCLUSIVE PRICE
    51.90 €
    38.40 € / 50 pairs
    Save 13.50 €
    26% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Latex, powder-free surgical gloves that provide everyday comfort, dexterity and reliable protection.
  • Proprietary anatomic-shaped glove formers to provide a comfortable fit and minimizes hand fatigue
  • Soft, malleable latex formulation with optimized thickness to provide good tactile sensitivity and dexterity
  • Polymer coated for fast and easy donning
Specifications
AQL 0.65
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Material Natural Rubber Latex
Powdered Powder Free
Size 8
Sterility Sterile