Learn More
Fisherbrand™ Media Bottle PET With Cap
Product Code. 30690123 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 24
Pack of 48
Pack of 60
Pack of 96
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30690123 1000 mL Pack of 24
30690122 125 mL Pack of 96
30690125 250 mL Pack of 60
30690124 500 mL Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30690123
In Stock
Add to basket
Edge
Add to basket
Made from high quality PET and the ultimate storage solution for media, biological buffers, and a wide range of reagents
  • Fisherbrand Media Bottles - These bottles are engineered to provide a comprehensive solution for storing media, biological buffers and a variety of reagents
  • These bottles are designed to be leak-proof, shatter proof and freezer-safe, making them ideal for storing and transporting valuable samples
  • Fisherbrand Media Bottles are the perfect choice for a reliable and convenient storage solution that meets the rigorous demands of modern laboratories
  • Fisherbrand's media bottles are the perfect blend of durability, safety and versatility
Specifications
Capacity (Metric) 1000 mL
Capacity (English) 0.264 gal.
Color Clear
Material PET
Sterility Sterile
Closure Included Yes
Disposable Single-use
Graduated Yes
Mouth Threaded
Shape Square
Fit for Purpose Sterile
Product Type Media Bottle
Closure Material HDPE
Autoclavable Not Autoclavable