missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Media Bottle PET With Cap

Product Code. 30690124 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 24
Pack of 48
Pack of 60
Pack of 96
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30690124 500 mL Pack of 48
30690123 1000 mL Pack of 24
30690122 125 mL Pack of 96
30690125 250 mL Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30690124
  • ONLINE EXCLUSIVE PRICE
    93.80 €
    84.15 € / Pack of 48
    Save 9.65 €
    10% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Made from high quality PET and the ultimate storage solution for media, biological buffers, and a wide range of reagents

  • Fisherbrand Media Bottles - These bottles are engineered to provide a comprehensive solution for storing media, biological buffers and a variety of reagents
  • These bottles are designed to be leak-proof, shatter proof and freezer-safe, making them ideal for storing and transporting valuable samples
  • Fisherbrand Media Bottles are the perfect choice for a reliable and convenient storage solution that meets the rigorous demands of modern laboratories
  • Fisherbrand's media bottles are the perfect blend of durability, safety and versatility

Specifications

Capacity (Metric) 500 mL
Capacity (English) 0.132 gal.
Color Clear
Material PET
Sterility Sterile
Closure Included Yes
Disposable Single-use
Graduated Yes
Mouth Threaded
Shape Square
Fit for Purpose Sterile
Product Type Media Bottle
Closure Material HDPE
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.