missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Media Supply Base Set

Product Code. 12671845
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12671845 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12671845 Supplier Hirschmann™ Supplier No. 9317000
  • ONLINE EXCLUSIVE PRICE
    460.95 €
    299.88 € / Pack of 1
    Save 161.07 €
    34% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enables suctioning of the medium directly from a separate storage container using a tube.

  • Accessory item
  • For use in titration
  • Provides base support

Specifications

Color White
Type Base Support
For Use With (Application) Enables suctioning of mediums
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.