missing translation for 'onlineSavingsMsg'
Learn More

Medicagenic Acid, TRC

Product Code. 30469807
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469807 1 mg 1mg
30468732 5 mg 5mg
30468861 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469807 Supplier Toronto Research Chemicals Supplier No. TRCM2038101MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-07-5
Molecular Formula C30H46O6
Molecular Weight (g/mol) 502.68
Synonym (2beta,3beta,4alpha)-2,3-dihydroxy-Olean-12-ene-23,28-dioic acid, 2beta,3beta-dihydroxy-Olean-12-ene-23,28-dioic acid, (2beta,3beta,4alpha)-2,3-Dihydroxyolean-12-ene-23,28-dioic acid, 2beta,3beta-Dihydroxyolean-12-en-23,28-dioic acid, Castanogenin, Medicogenic acid, NSC 382024
IUPAC Name (2S,3R,4S,4aR,6aR,6bS,8aS,12aS,14aR,14bR)-2,3-dihydroxy-4,6a,6b,11,11,14b-hexamethyl-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicene-4,8a-dicarboxylic acid
SMILES CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@H](O)[C@H](O)[C@](C)([C@@H]5CC[C@@]34C)C(=O)O)[C@@H]2C1)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C30H46O6/c1-25(2)11-13-30(24(35)36)14-12-27(4)17(18(30)15-25)7-8-20-26(3)16-19(31)22(32)29(6,23(33)34)21(26)9-10-28(20,27)5/h7,18-22,31-32H,8-16H2,1-6H3,(H,33,34)(H,35,36)/t18-,19-,20+,21+,22-,26+,27+,28+,29-,30-/m0/s1
Formula Weight 502.3294
Chemical Name or Material Medicagenic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.