missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Spark-Free interior, Lab Fridge

Product Code. 15230469 Shop All Liebherr Appliances Products
Change view
Click to view available options
Capacity (Metric):
333 L
554 L
Style:
Tall
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Style missing translation for 'unitSize'
15230469 554 L Tall Each
15202596 333 L Tall Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15230469 missing translation for 'mfr' Liebherr™ missing translation for 'supplierNo' Lkexv 5400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For secure, accurate and long-lasting, spark-free interior, fridge storage of non temperature-sensitive products, the Liebherr™ Spark-Free Interior, Lab Fridge is designed for general purpose applications with Liebherr build-quality you can rely on.

  • Spark-Free Lab Fridge for storage of solvents and reagents, reach-in, 1-section, width 600mm.
  • Temperature range +1 / +15°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration.
  • Climate class 4, refrigerant R 600a, connection rating 1,0A, 230v/50/1-ph,energy consumption 328 kWh/a
  • This model is intended for the spark-free storage of chemicals where a high level of temperature stability is not required
  • ATEX-certified spark-free interior
  • Analogue controls with digital temperature display in the door.
  • Door lock, reversible hinges
  • 40 kg shelf load

Specifications

Type Laboratory Refrigerator
Capacity (Metric) 554 L
Temperature Range 1°C to 15°C
Temperature Resolution 1.0°C
Temperature Stability Gradient 6,9°C
Refrigerant R 600a
Door Style Solid
No. of Doors 1
Hinge Side Right, reversible
No. of Drawers 0
Shelves 5
Capacity (Metric) Shelf 40 kg
Cabinet Material Steel
Exterior Finish White-coated Steel
Insulation 40 mm HDPE
Interior Polystyrol
Certifications/Compliance ATEX Certified
Style Tall
Display Digital, External
Casters No
Color White
Control Type Analog
Defrost Automatic
Compressor Forced-air
Depth (Metric) Exterior 73 cm
Depth (Metric) Interior 56 cm
Height (Metric) Exterior 164 cm
Height (Metric) Interior 145.2 cm
Width (Metric) Exterior 75 cm
Width (Metric) Interior 60 cm
Amperage 1.5 A
Voltage 220/240 V
Electrical Requirements 220 - 240 V 50 Hz 1.5 A
Frequency 50 Hz
Energy Usage 359 kWh/a
Shipping Weight (Metric) 84 kg
Weight (Metric) 77 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.