Learn More
Manutan Medium duty alloy track, 40 roller bearings, P50, 500mm width
Product Code. 12831998
Encompass_Preferred
Product Code. 12831998 Supplier Manutan Supplier No. A341798
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Lightweight aluminum gravity conveyor track with PVC roller bearings and mild steel bearings. Frame: 63mm x 25mm x 3mm aluminum channel. 40/50mm diameter PVC roller bearings. 30kg load capacity per metre.
  • 40mm diameter roller bearing with mild steel bearings and spindles
  • Track width: 500mm
  • Roller bearing pitch: 50mm