Learn More
Manutan Medium duty shelving, Rapid 2, blue/grey, three shelves, 990mm x 915mm x 610mm
Product Code. 12827656
Encompass_Preferred
Product Code. 12827656 Supplier Manutan Supplier No. A360955
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A versatile shelving system used for hand loaded applications. Shelf load capacity of 200kg-340kg. Simple boltless assembly - frames and beams simply clip together and only require a rubber mallet. Shelves adjustable by 38mm increments, even when built. High density chipboard shelves with max shelf capacity of up to 340kg. 2mm European structural quality steel. Hardwearing powder coated finish. Load capacity TUV certified - test to establish safe working loads. Supplied flat-packed.
  • Blue/grey
  • Three shelves
  • Shelf capacity UDL: 340kg
  • Dimensions (H x W x D): 990mm x 915mm x 610mm