missing translation for 'onlineSavingsMsg'
Learn More

CHEMGENE MedLab Multi-Surface Disinfectant Cleaner

Product Code. 30245733 Shop All Chemgene Products
Change view
Click to view available options
Color:
Blue
Translucent Yellow
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30245733 Translucent Yellow Pack of 6
30245734 Blue Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30245733 Supplier CHEMGENE Supplier No. SKU093A
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use hard surface disinfectant for laboratory, critical services, and forensics settings.

  • Next-generation CHEMGENE HLD4L and HLD4H, CHEMGENE MedLab is a range of surface disinfectants created specifically to meet the demands of laboratory environments.
  • CHEMGENE MedLab Multi-Surface Disinfectants are proven to be effective under the following microbiological test conditions; Bactericidal: EN 17387, EN 13727, EN 16615. Legionella: EN 13623. Yeasticidal: EN 13624, EN 17387, EN 16615. Fungicidal: EN 17387, EN 13624. Virucidal: EN 14476, EN 16777 (Norovirus, Adenovirus). Bacteriophage: EN 13610.
  • Fast-acting efficacy from as little as 60 seconds
  • Denatures DNA/RNA
  • Biodegradable formulation
  • Available in eucalyptus scent or un-fragranced
  • Manufactured in the UK to ISO 9001 quality standards
  • Non-corrosive
  • Alcohol-free
  • No rinsing required
  • Compatible with a wide range of materials including mild steel, stainless steel, copper, aluminium, acetal, nylon and rubber.
  • Bottle made from 30% recycled HDPE
  • Bottle and trigger recyclable after use

Specifications

Product Type Foaming Trigger Sprayer
Color-coded No
Form Clear Liquid
pH 10.0 to 11.0
Scent Unscented
Sterility Non-sterile
Appearance Clear Pale Yellow
Autoclavable Autoclavable
Color Translucent Yellow
Concentration 10%
Container Type Spray Bottle - 30% Recycled HDPE
Content And Storage Store out of direct sunlight.
Dilution Product does not require dilution
Dimensions (L x W) 63 x 110 mm
Dimensions (L x W x H) 63 x 110 x 287 mm
Disposable Single-use
Filtered Filtered
Flammable No
Format Trigger Spray
For Use With (Application) Cleaning and disinfection of hard surfaces and equipment
Height (English) 11.2 in.
Height (Metric) 287 mm
Length (English) 2.4 in.
Length (Metric) 63 mm
Packaging Trigger Spray - Bottle 30% Recycled HDPE
Size 750 mL
Volume (Metric) 750 mL
Weight (English) 28.5 oz.
Weight (Metric) 809 g
Width (English) 4.3 in.
Width (Metric) 110 mm
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • MedLab RTU Spray Multi-Surface Disinfectant
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Didecyldimethylammonium chloride, Alkyl(C12-16) dimethylbenzyl ammonium chloride, N-(3-Aminopropyl)

Use biocides safely. Always read the product label and information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.