missing translation for 'onlineSavingsMsg'
Learn More

CHEMGENE MedLab Multi-Surface Disinfectant Cleaner

Product Code. 30245734 Shop All Chemgene Products
Change view
Click to view available options
Color:
Blue
Translucent Yellow
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30245734 Blue Pack of 6
30245733 Translucent Yellow Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30245734 Supplier CHEMGENE Supplier No. SKU095A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use hard surface disinfectant for laboratory, critical services, and forensics settings.

  • Next-generation CHEMGENE HLD4L and HLD4H, CHEMGENE MedLab is a range of surface disinfectants created specifically to meet the demands of laboratory environments.
  • CHEMGENE MedLab Multi-Surface Disinfectants are proven to be effective under the following microbiological test conditions; Bactericidal: EN 17387, EN 13727, EN 16615. Legionella: EN 13623. Yeasticidal: EN 13624, EN 17387, EN 16615. Fungicidal: EN 17387, EN 13624. Virucidal: EN 14476, EN 16777 (Norovirus, Adenovirus). Bacteriophage: EN 13610.
  • Fast-acting efficacy from as little as 60 seconds
  • Denatures DNA/RNA
  • Biodegradable formulation
  • Available in eucalyptus scent or un-fragranced
  • Manufactured in the UK to ISO 9001 quality standards
  • Non-corrosive
  • Alcohol-free
  • No rinsing required
  • Compatible with a wide range of materials including mild steel, stainless steel, copper, aluminium, acetal, nylon and rubber.
  • Bottle made from 30% recycled HDPE
  • Bottle and trigger recyclable after use

Specifications

Product Type Foaming Trigger Sprayer
Color-coded Yes
Form Liquid Spray
pH 10.0 to 11.0
Scent Eucalyptus
Sterility Non-sterile
Appearance Light Blue
Autoclavable Autoclavable
Color Blue
Concentration 10%
Container Type Trigger Spray - 30% rRcycled HDPE
Content And Storage Store out of direct sunlight.
Dilution Product does not require dilution
Dimensions (L x W) 63 x 110 mm
Dimensions (L x W x H) 63 x 110 x 287 mm
Disposable Single-use
Filtered Filtered
Flammable No
Format Trigger Spray
For Use With (Application) Cleaning and disinfection of hard surfaces and equipment
Height (English) 11.2 in.
Height (Metric) 287mm
Length (English) 2.4 in.
Length (Metric) 63 mm
Packaging Trigger Spray - Bottle 30% Recycled HDPE
Size 750 mL
Volume (Metric) 750 mL
Weight (English) 28.6 oz.
Weight (Metric) 819 g
Width (English) 4.3 in.
Width (Metric) 110 mm
Show More Show Less
Product Identifier
  • Chemgene
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Didecyldimethylammonium chloride

Use biocides safely. Always read the product label and information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.