Learn More
Chemgene MedLab Multi-Surface Disinfectant Concentrate
Description
CHEMGENE MEDLAB Multi-Surface Disinfectants are suitable in a variety of applications within laboratory, critical or emergency services, government agencies, quality control laboratries, and healthcare environments. Intended for use on surfaces and equipment including benches, shelves, fridges, water baths, centrifuges, incubators, discard jars, pipettes and many more, CHEMGENE MEDLAB Multi-Surface Disinfectants are versatile and ideal for implementing effective biosecurity measures in a wide range of organizations.
Suitable for use and compatible with hard steel, mild steel, copper, aluminium, rubber, plastics, acetal and nylon, CHEMGENE MEDLAB Multi-Surface Disinfectants are non-corrosive in use and do not leave problematic residues on surfaces after use which may interfere with validated assays or other laboratory work.
This new generation of CHEMGENE, developed by the hygiene specialists at Byotrol, benefits from an optimized formulation, which is biodegradable and is supported by a robust data set. The improved balance of efficacy with active material concentration has resulted in a superior disinfectant which is exceptionally effective on a wide range of pathogenic microorganisms.
CHEMGENE’s broad spectrum efficacy against bacteria, yeasts, viruses and fungi, has been demonstrated by a robust data set performed to the following European PT2 test standards, including challenging medical standards performed in dirty conditions
EN 13727 (bactericidal)
EN 17387 (bactericidal)
EN 13624 (yeasticidal, fungicidal)
EN 17387 (yeasticidal, fungicidal)
EN 16777 (virucidal – all enveloped viruses)
EN 16777 (Adenovirus, Norovirus)
EN 14476 (virucidal – all enveloped viruses)
EN 14476 (Adenovirus, Norovirus)
EN 13623 (Legionella)
EN 13610 (bacteriophage)
CHEMGENE MEDLAB Multi-Surface Disinfectants are proven effective against:
Norovirus
Adenovirus
Coronavirus
Herpes Simplex Virus
Respiratory syncytial virus
Hepatitis B & C
HIV
Candida auris
Enterococcus faecium
Klebsiella pneumoniae
Staphylococcus aureus
Acinetobacter baumannii
Pseudomonas aeruginosa
Enterobacter spp.
Streptococcus pyogenes
Moraxella catarrhalis
Aspergillus brasiliensis
Legionella
E.coli
Epstein Barr Virus
Cytomegalovirus
Human herpesvirus 6
Zika virus
Varicella zoster virus
Parainfluenza
Influenza
Human T-cell lymphotropic virus
Validated testing performed by the University of Chester confirms that CHEMGENE MEDLAB Multi-Surface Disinfectants denatures DNA in 5 minutes when used at 1% dilution in low soil.
Easy-to-dilute and simple to use, CHEMGENE MEDLAB Multi-Surface Disinfectant can be diluted to 1% for general cleaning and disinfection and diluted to 10% for the high-risk disinfection of bodily fluid spillages, or the disinfection of liquid waste. The 1 litre self-dosing format has volume markers to aid consistently correct dilution of the concentrate and a helpful dilution poster is available as a visual reminder.
Once diluted by the end-user, CHEMGENE is effective for up to three months. Reusable 30% recycled plastic trigger spray bottles and color-coded dilution labels are also available to aid and improve compliance and reduce the impact on our environment.
CHEMGENE MEDLAB products are compliant with all relevant regulations in the UK including CLP, REACH and the Biocidal Products Regulation (BPR) including Article 95.
CHEMGENE MEDLAB products are not intended for the disinfection of medical devices. Use biocides safely. Always read the label and product information before use.
Specifications
Specifications
| Product Type | Disinfectant Concentrate |
| Color-coded | No |
| Form | Concentrated Liquid |
| pH | 10.5-11.4 |
| Scent | Eucalyptus |
| Sterility | Non-sterile |
| Appearance | Blue |
| Color | Blue |
| Disposable | Single-use |
| Filtered | Non-filtered |
| Show More |
Safety and Handling
- CHEMGENE MedLab Multi-Surface Disinfectant
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P264-Wash contaminated skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Didecyldimethylammonium chloride, Limonene
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.