missing translation for 'onlineSavingsMsg'
Learn More

Medronic Acid, TRC

Product Code. 30135627
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135627 50 mg 50mg
30008426 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135627 Supplier Toronto Research Chemicals Supplier No. TRCM20350050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1984-15-2
Molecular Formula C H6 O6 P2
Molecular Weight (g/mol) 176
Synonym Medronic acid, Methylenediphosphonic acid, P,P'-Methylenebisphosphonic acid, Phosphonic acid, methylenebis- (9CI), Phosphonic acid, methylenedi- (6CI,7CI,8CI), MDP, Medronate, Medronic acid, Methanebisphosphonic acid, Methanediphosphonic acid, Methylene-1,1-bisphosphonic acid, Methylenebis[phosphonic acid], Methylenediphosphonic acid
IUPAC Name phosphonomethylphosphonic acid
SMILES OP(=O)(O)CP(=O)(O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/CH6O6P2/c2-8(3,4)1-9(5,6)7/h1H2,(H2,2,3,4)(H2,5,6,7)
Formula Weight 175.964
Chemical Name or Material Medronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.