missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mefenamic-d4 Acid (benzoic-3,4,5,6-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Mefenamic-d4 Acid |
| CAS | 1216745-79-7 |
| Molecular Formula | C15 D4 H11 N O2 |
| Quantity | 0.01 g |
| Synonym | Benzoic-2,3,4,5-d4 acid, 6-[(2,3-dimethylphenyl)amino]- (ACI), 6-[(2,3-Dimethylphenyl)amino]benzoic-2,3,4,5-d4 acid (ACI), Mefenamic-d4 Acid, Mefenamic Acid D4 (benzoic acid 3,4,5,6-D4) |
| InChI Formula | InChI=1 S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16 H,1-2H3,(H,17,18)/i3D,4 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(Nc2cccc(C)c2C)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-(2,3-dimethylanilino)benzoic acid |
| Molecular Weight (g/mol) | 245.3097 |
| Formula Weight | 245.1354 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction