missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mefloquine hydrochloride

Product Code. 18777656
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18777656 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18777656 Supplier R&D Systems™ Supplier No. 6819/50
  • ONLINE EXCLUSIVE PRICE
    91.05 €
    81.90 € / 50mg
    Save 9.15 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cx36 and Cx50 gap channel blocker

Mefloquine hydrochloride is a Cx36 and Cx50 gap channel blocker (IC50 values are 0.3 and 1.1 μM, resepctively). Blocks gap junctional-coupling between interneurons in neocortical slices. Also antimalarial. Bind 80S ribosome of Plasmodium falciparum to inhibit protein synthesis. Improves survival in P berghei-infected mice. Additionally exhibits antischistosomal activity in vitro and in vivo. Exhibits antiviral activities against SARS-CoV-2 (IC50 < 10 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (αS)-rel-α-(2R)-2-Piperidinyl-2,3-bis(trifluoromethyl-4-quinolinemethanol hydrochloride
CAS 51773-92-3
Quantity 50 mg
Product Line Tocris
Molecular Formula C17H16F6N2O.HCl
Formula Weight 414.77
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.