Learn More
STEMCELL Technologies MegaCult™-C Avidin-Alkaline Phosphatase Conjugate
Description
- MegaCult™-C Staining Kit for CFU-Mk contains all the reagents needed to stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium
- Megakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system
- The kit is also suitable for detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk)
Specifications
Specifications
| Quantity | 200 μL |
| Product Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Colony Assay, Immunocytochemistry |