missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Megaplex™ PreAmp Primers, Human Pool Set v3.0
GREENER_CHOICE

Product Code. 11680089 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 reactions
Unit Size:
50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11680089 50 reactions 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11680089 Supplier Applied Biosystems™ Supplier No. 4444748
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Megaplex™ PreAmp Primers, Human Pool Set v3.0

For use when assay sensitivity of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. With content matched to the Megaplex™ RT Primers, Megaplex™ PreAmp Primers, Human Pool Set v3.0 is used with Megaplex™ RT Primers, Human Pool Set v3.0. Following reverse transcription using Megaplex™ RT Primers, Megaplex™ PreAmp Primers along with the TaqMan™ PreAmp Master Mix uniformly amplify all microRNAs prior to quantitation by real-time PCR using TaqMan™ Array MicroRNA Cards.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C
GC-Rich PCR Performance Low
PCR Method qPCR
Primer PreAmp Primers
Product Line Megaplex
Product Type Preamp Primer
Quantity 50 reactions
Species Human
Sufficient For 50 Reactions
No. of Reactions 50 reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.