missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAscript™ RNAi Kit

Product Code. 10536935
Change view
Click to view available options
Quantity:
20 reactions
Unit Size:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10536935 20 reactions 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10536935 Supplier Invitrogen™ Supplier No. AM1626
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEGAscript™ RNAi Kit

The Invitrogen™ MEGAscript™ RNAi Kit is for the synthesis of large mass amounts of dsRNA for RNAi experiments in nonmammalian systems. The MEGAscript RNAi Kit uses Invitrogen's patented high-yield transcription technology and includes reagents for transcription, nuclease digestion, and clean up. The kit includes sufficient reagents for 20 reactions.

Synthesize Large Amounts of dsRNA:
The ability to generate large amounts of clean, double-stranded RNA is essential for the success of RNAi experiments. The MEGAscript RNAi Kit allows RNAi researchers to synthesize 50 to 100μg or more of double-stranded RNA in a single transcription reaction. The yield is typically 10–50 times the amount of RNA produced by conventional transcription reactions. DNA of either PCR or plasmid origin can be used as a template for the transcription reaction. The sense and antisense strands can be synthesized from a PCR-generated DNA template containing T7 RNA Polymerase promoters on both ends of the template, eliminating the need for a separate annealing step.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage T7 Enzyme Mix, 10X T7 Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, 10X Binding Buffer, Elution Solution, RNase, DNase I, 10X Digestion Buffer, 2X Wash Solution, and Control Template are all stored at –20°C, Nuclease-free Water may be store at any temperature. Filter Cartridges and Collection Tubes are stored at room temperature.
Control Type Transcription Control
Sample Type Double-Stranded RNA (Long)
No. of Reactions 20 reactions
Product Line Ambion, MEGAscript
Quantity 20 reactions
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.