missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The sporty, ergonomic design and ultra-comfortable fit means these goggles both protect and allow you to get on with your day.
General features
- Functional goggles in ski goggle design with panoramic view
- Unique, frameless lens design with maximum optimised field of view for better hazard perception and increased safety
- Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
- Sustainable benefit: headband can be replaced if necessary
- Metal-free
Protection features
- Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
- UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
- Increased mechanical strength – BT marking (120 m/s; tested under extreme temperatures -5°C, +55°C)
Comfort features
- Soft, adaptive face pad provides a universal fit for different head and face shapes and a tight fit without pressure points
- Generous internal volume and recesses for the side arms of the glasses allow comfortable wear without pressure points, even for glasses wearers
- Indirect ventilation system increases air circulation
- 30 mm wide length adjustable fabric headband for optimum hold and individual adjustment
Specifica
Specifica
| Lens Type | Infradur Plus |
| Lens Coating | Anti-fog |
| Certifications/Compliance (Europe) | EN 166, EN 169 |
| Lens Tint | Welding Shade 5, Gray |
| Product Type | Welding Spectacles |
| Frame Color | Black |
| Includes | Textile Headband |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?