missing translation for 'onlineSavingsMsg'
Learn More

Melamine, TRC

Product Code. 30137222
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137222 5 g 5g
17944555 1 g 1g
17982545 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137222 Supplier Toronto Research Chemicals Supplier No. TRCM2087005G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-78-1
Molecular Formula C3 H6 N6
Molecular Weight (g/mol) 126.12
Synonym 1,3,5-Triazine-2,4,6-triamine, Metformin Hydrochloride Imp. D (EP), Melamine
IUPAC Name 1,3,5-triazine-2,4,6-triamine
SMILES Nc1nc(N)nc(N)n1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)
Formula Weight 126.0654
Percent Purity >95%
Chemical Name or Material Melamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.