missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Melanocyte-Stimulating Hormone-Release Inhibiting Factor, synthetic

Product Code. 15450117
Change view
Click to view available options
Quantity:
250mg
1g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450117 250mg 250mg
15440117 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450117 Supplier Thermo Scientific Chemicals Supplier No. J60045.MD
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis. | 284.36 g/mol

Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 284.36
Quantity 250mg
Packaging Glass bottle
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.