Learn More
DiscoverX Melanotan II
Product Code. 17382449
Encompass
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
17382449 1 mg 1mg
1 options
Product Code. 17382449 Supplier DiscoverX Supplier No. 921122
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Melanotan II is an agonist for MC1R, MC3R, MC4R and MC5R. Provided as 1mg of dry powder alongside reconstitution buffer.
Melanotan II is a synthetic analog of the melanocortin peptide hormone α-MSH. Effects of its use include melanogenesis and aphrodesia. Clinical development ceased in 2003 with no products FDA approved, but melanotan II products are currently sold and purchased online. Acts as an agonist for MC1R, MC3R, MC4R, and MC5R. Provided as 1mg of dry powder alongside reconstitution buffer.
Specifications
Product Type Control Ligands
Quantity 1 mg