missing translation for 'onlineSavingsMsg'
Learn More

Melinamide, MedChemExpress

Product Code. 16493528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493528 10 mg 10mg
16436257 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16493528 Supplier Medchem Express Supplier No. HY101722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Melinamide, an amide derivative of an unsaturated long-chain fatty acid, is an inhibitor of cholesterol absorption with an IC50 of 20.9 μM.

Melinamide, an amide derivative of an unsaturated long-chain fatty acid, is an inhibitor of cholesterol absorption with an IC50 of 20.9 μM.

Specifications

Chemical Name or Material Melinamide
CAS 14417-88-0
Color Colorless
Purity Grade Notes Research
Molecular Formula C26H41NO
Quantity 10 mg
Synonym AC 223 DL-N-(α-Methylbenzyl)linoleamide
Solubility Information DMSO : 200 mg/mL (521.36 mM; Need ultrasonic)
SMILES CCCCC/C=C\C/C=C\CCCCCCCC(NC(C1=CC=CC=C1)C)=O
Molecular Weight (g/mol) 383.61
Formula Weight 383.61
Percent Purity 99.94%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.