missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Meloxicam, 99-101%

Product Code. 15569483 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15569483 25 g 25g
15559483 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15569483 Supplier Thermo Scientific Chemicals Supplier No. 459550250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71125-38-7
Molecular Formula C14H13N3O4S2
Molecular Weight (g/mol) 351.40
MDL Number MFCD00868752
InChI Key DWMREKMVXIFPFM-ACCUITESSA-N
Synonym meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin
PubChem CID 54677470
ChEBI CHEBI:6741
IUPAC Name 4-hydroxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-1$l^{6},2-benzothiazine-3-carboxamide
SMILES CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Melting Point 255.0°C
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (105°C, 4 hr)
Infrared Spectrum Authentic
Quantity 25 g
Solubility Information Solubility in water: 7,5g/L. Other solubilities: 5g/L dmso
Formula Weight 351.41
Percent Purity 99 to 101%
Chemical Name or Material Meloxicam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.