missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam

Product Code. 15483369
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483369 25 g 25g
15473369 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483369 Supplier Thermo Scientific Alfa Aesar Supplier No. J60635.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol

Meloxicam is developed for the treatment of rheumatoid arthritis and osteoarthritis. It is currently for the treatment of osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (22 mg/ml), DMSO (25 mg/ml), DMF, acetone(slightly soluble), ethanol(slightly soluble), and methanol(slightly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71125-38-7
Molecular Formula C14H13N3O4S2
Molecular Weight (g/mol) 351.40
MDL Number MFCD00868752
InChI Key DWMREKMVXIFPFM-ACCUITESSA-N
Synonym meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin
PubChem CID 54677470
ChEBI CHEBI:6741
IUPAC Name (3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
SMILES CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Melting Point 255°C
Color Yellow
Quantity 25 g
UN Number UN2811
Merck Index 14,5826
Solubility Information Soluble in water (22mg/ml),DMSO (25mg/ml),DMF,acetone(slightly soluble),ethanol(slightly soluble),and methanol(slightly soluble).
Formula Weight 351.39
Physical Form Powder
Chemical Name or Material Meloxicam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.