missing translation for 'onlineSavingsMsg'
Learn More

Melphalan, TRC

Product Code. 30468860
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30468860

Brand: Toronto Research Chemicals TRCM21690050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Melphalan
CAS 148-82-3
Molecular Formula C13 H18 Cl2 N2 O2
Quantity 50 mg
Synonym Melphalan, 4-[Bis(2-chloroethyl)amino]-l-phenylalanine, L-Phenylalanine, 4-[bis(2-chloroethyl)amino]-, Alanine, 3-[p-[bis(2-chloroethyl)amino]phenyl]-, L- (8CI), 3025CB, 4-Bis(2-chloroethyl)amino-L-phenylalanine, 67: PN: WO2010011890 PAGE: 37 claimed protein, Alanine nitrogen mustard, Alkeran, CB 3025, L-PAM, L-Phenylalanine mustard, L-Phenylalanine mustard hydrochloride, L-Sarcolysin, L-Sarcolysine, L-Sarkolysin, Levofalan, Levofolan, Levopholan, Melfalan, Melphalan, NSC 241286, NSC 8806, Phenylalanine mustard, Sarcoclorin
InChI Formula InChI=1S/C13H18Cl2N2O2/c14-5-7-17(8-6-15)11-3-1-10(2-4-11)9-12(16)13(18)19/h1-4,12H,5-9,16H2,(H,18,19)/t12-/m0/s1
SMILES N[C@@H](Cc1ccc(cc1)N(CCCl)CCCl)C(=O)O
IUPAC Name (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid
Molecular Weight (g/mol) 305.2
Formula Weight 304.0745
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.