missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Membrane Permeability Dead Cell Apoptosis Kit with PO-PRO™-1 and 7-Aminoactinomycin D, for flow cytometry

Product Code. 10338452
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10338452 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10338452 Supplier Molecular Probes™ Supplier No. V35123
  • ONLINE EXCLUSIVE PRICE
    676.00 €
    479.96 € / 1 sets
    Save 196.04 €
    29% Off
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Vybrant™ Apoptosis Assay Kit #13 *PO-PRO™-1/7-aminoactinomycin D* *for flow cytometry* *200 assays*

This flow cytometry product detects apoptosis on the basis of changes that occur in the permeability of cell membranes using PO-PRO™-1 and 7-aminoactinomycin D (7-AAD) nucleic acid stains. After staining a cell population with PO-PRO™-1 dye and 7-AAD, apoptotic cells show violet fluorescence, dead cells show violet and red fluorescence, and live cells show little or no fluorescence. These populations can easily be distinguished by a flow cytometer that uses both a violet laser and the 488 nm line of an argon-ion laser for excitation. Each kit contains sufficient reagents for approximately 200 flow cytometry tests.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Format Tube, Slide
Product Type Dead Cell Apoptosis Kit
Content And Storage Contains 1 vial of PO-PRO™-1 dye (500 μL), and 1 vial of 7-AAD (7-aminoactinomycin D, 200 μL).

Store in refrigerator (2–8°C) and protect from light.
Flow Cytometer Laser Lines 405, 488
Excitation/Emission PO-PRO™-1: 434⁄456, 7-AAD: 546⁄647
No. of Reactions 200
Shipping Condition Room Temperature
Product Line Vybrant
For Use With (Application) Flow Cytometry
Conjugate 7-AAD, PO-PRO-1
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.