Description
- For 5 - 10 L canisters in the dispensing drawer
- The suction lance features level detection
- Installed by Customer Service
- Extra disp. canisters with installation by customer service
Specifications
Specifications
| Product Type | Dispensing System |
| Model | APLW 884 |
| For Use With (Equipment) | Washer-Disinfectors |
| Includes | 5 to 10 L Canister |
| Quantity | 1 Ea. |