missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MembranePro™ Functional Protein Expression Kit

Product Code. 17173029
Change view
Click to view available options
Quantity:
10 reactions kit
Unit Size:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17173029 10 reactions kit 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17173029 Supplier Invitrogen™ Supplier No. A11667
  • ONLINE EXCLUSIVE PRICE
    2775.00 €
    2275.50 € / 10 reactions
    Save 499.50 €
    18% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

MembranePro™ Functional Protein Expression Kit - 10rxn

For customers studying GPCRs and other membrane proteins, MembranePro™ is the ready-to-use system that delivers enriched, functional membrane proteins efficiently and reliably. Benefits of MembranePro™ include:

  • Proteins are displayed on human (or other mammalian) cell membranes
  • Cellular quality control and mammalian posttranslational processing helps produce functional protein
  • Proteins bud off as lipoparticles and are easily collected from the culture medium
  • Lipoparticles are enriched with expressed receptors

    MembranePro™ workflow is less labor intensive in comparison to traditional production of membrane fractions. Briefly, the MembranePro™ protocol allows you to:

    1. Clone your gene of interest using a TOPO™ vector
    2. Transfect with Lipofectamine™ 2000 and MembranePro™ Reagent into 293FT cells
    3. Harvest lipoparticles 48h post-transfection, concentrate with MembranePro™ Precipitation Mix
    4. Store lipoparticles at -80 C or proceed to downstream assays

    MembranePro™ products are offered in two configurations – as an expression kit (TOPO™ vector, Lipofectamine™ 2000, MembranePro™ Precipitation Mix, MembranePro™ Reagent, 293FT cells) and as a support kit (Lipofectamine™ 2000, MembranePro™ Precipitation Mix, MembranePro™ Reagent). The expression kit is only available in a 10 reaction size; the support kit is available in 10, 60 and 600 reaction sizes.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Type MembranePro Functional Protein Expression Kit
    Delivery Type Virus-Like Particles (VLPs)
    Quantity 10 reactions kit
    Content And Storage The kit contains:
    •pEF6 V5-His TOPO™ TA Vector Kit for at least 10 TOPO™ cloning reactions (Store at -20°C)
    •3 x10^6 293FT Cells (Store in liquid nitrogen)
    •1 x 75 ml MembranePro™ Precipitation Mix (Store at room temperature)
    •1 x 0.3 mg (1 mg⁄ml) MembranePro™ Reagent (Store at -20°C)
    •3 x 0.75 ml Lipofectamine™ 2000 (Store at +4°C)
    •Manuals
    Selection Agent (Eukaryotic) Blasticidin
    Cell Line 293FT
    Expression Mechanism Cell-Based Expression
    High-throughput Compatibility High-throughput Compatible
    Selection Marker Eukaryotic BsdR
    Expression System Mammalian
    Expressed Protein Type Membrane Protein
    Product Line MembranePro
    For Use With (Application) Membrane Protein Expression
    Promoter EF-1α
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.