Learn More
Emma Safety Footwear Mendoza Safety Shoes
Product Code. 15908781
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15908781 37 1 pair
15918781 38 1 pair
15928781 39 1 pair
15938781 40 1 pair
15948781 41 1 pair
15958781 42 1 pair
15968781 43 1 pair
15978781 44 1 pair
15988781 45 1 pair
15998781 46 1 pair
15908791 47 1 pair
15918791 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15908781 Supplier Emma Safety Footwear Supplier No. MM58284837
  • ONLINE EXCLUSIVE PRICE
    238.35 €
    149.10 € / 1 pairs
    Save 89.25 €
    37% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
State-of-the art Sympatex™ construction provides perfect foot protection for hands-on roles within the Chemical Industry and Agriculture. Virtually waterproof, the Mendoza boots allow feet to breathe properly, keeping them cool and dry.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck / Sympatex™ construction
  • Waterproof
  • GripForce™ Duo Heat Resistant Outsole
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Brown
Design Mendoza
Liner Material Hydro-Tec™ Sanitized Silver / Sympatex™
Material Oiled Nubuck
Size 37
Toe Material Steel