Learn More
Emma Safety Footwear Mento Safety Shoes
Water repellant for up to 4 hours and ideally suited to working outdoors. Designed to offer exceptional all over foot protection with a reinforced steel plate in the sole, and strong, supportive leather uppers. A double-density polyurethane sole protects feet from dangers above and below.
Brand:  Emma Safety Footwear MM58154840
Additional Details : Weight : 2.03200kg
 View more versions of this product
Product Code. 15928771
  • 177.02 € / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
EN ISO 20345:2011
Mento
Leather
Steel
Brown
Hydro-Tec™ Sanitized Silver
40