missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mephenesin, TRC
Specifications
Specifications
| CAS | 59-47-2 |
| Molecular Formula | C10 H14 O3 |
| Quantity | 250 mg |
| Synonym | Mephenesin, 1,2-Propanediol, 3-(2-methylphenoxy)-, 1,2-Propanediol, 3-(o-tolyloxy)- (8CI), 3-(2-Methylphenoxy)-1,2-propanediol, (±)-Mephenesin, 1,2-Dihydroxy-3-(2-methylphenoxy)propane, 3-(2-Methylphenoxy)propane-1,2-diol, 3-(2-Tolyloxy)-1,2-propanediol, 3-(o-Methylphenoxy)-1,2-propanediol, 3-(o-Tolyloxy)-1,2-propanediol, 3-o-Toloxy-1,2-propanediol, Atensin, Avosyl, Avoxyl, BDH 312, Cresoxydiol, Cresoxypropanediol, Curythan, Daserd, Daserol, Decontractyl, Dioloxol, Glyceryl o-tolyl ether, Glykresin, Glyotol, Kinavosyl, Lissephen, Memphenesin, Mephedan, Mephelor, Mephenesine, Mephensin, Mepherol, Mephesin, Mephin, Mephson, Mervaldin, Myanesin, Myanol, Myasin, Mycocuran, Myodetensin, Myodetensine, Myolysin, Myopan, Myoserol, Myoten, Myoxane, NSC 25234, NSC 36140, NSC 50788, NSC 8134, Oranixon, Prolax, RP 3602, Relaxar, Relaxil, Renarcol, Rhex, Sansdolor, Sinan, Spartoloxyn, Spasmolyn, Stilalgin, Thoxidil, Tolansin, Tolax, Tolcil, Tolhart, Tolosate, Toloxyn, Tolserol, Tolseron, Tolsin, Tolulexin, Tolulox, Tolyspaz, Walconesin, o-Cresyl glycerol ether, o-Cresyl α-glyceryl ether |
| InChI Formula | InChI=1S/C10H14O3/c1-8-4-2-3-5-10(8)13-7-9(12)6-11/h2-5,9,11-12H,6-7H2,1H3 |
| SMILES | Cc1ccccc1OCC(O)CO |
| IUPAC Name | 3-(2-methylphenoxy)propane-1,2-diol |
| Molecular Weight (g/mol) | 182.22 |
| Formula Weight | 182.0943 |
| Chemical Name or Material | Mephenesin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?