missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mephenytoin

Product Code. 17201272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
17201272 100 mg 100mg
17291262 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 17201272

Brand: Thermo Scientific Chemicals 466691000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-12-4
Molecular Formula C12H14N2O2

Specifications

Quantity 100 mg
Solubility Solubility aq. soln.: insoluble.
Melting Point 135.0°C to 137.0°C
Formula Weight 218.25
InChI Key GMHKMTDVRCWUDX-UHFFFAOYNA-N
SMILES CCC1(NC(=O)N(C)C1=O)C1=CC=CC=C1
IUPAC Name 5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
Molecular Weight (g/mol) 218.26
Chemical Name or Material Mephenytoin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.