missing translation for 'onlineSavingsMsg'
Learn More

Meprednisone 17,21-Dipropionate-d10, TRC

Product Code. 30477542
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477542 10 mg 10mg
30473219 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477542 Supplier Toronto Research Chemicals Supplier No. TRCM22505210MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Meprednisone 17,21-Dipropionate-d10
Molecular Formula C28 D10 H26 O7
Quantity 10 mg
Synonym (16beta)-16-methyl-17,21-bis(1-oxopropoxy-d10)pregna-1,4-diene-3,11,20-trione
InChI Formula InChI=1S/C28H36O7/c1-6-23(32)34-15-22(31)28(35-24(33)7-2)16(3)12-20-19-9-8-17-13-18(29)10-11-26(17,4)25(19)21(30)14-27(20,28)5/h10-11,13,16,19-20,25H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,25+,26-,27-,28-/m0/s1/i1D3,2D3,6D2,7D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C(=O)OCC(=O)[C@@]1(OC(=O)C([2H])([2H])C([2H])([2H])[2H])[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC Name [2-oxo-2-[(8S,9S,10R,13S,14S,16S,17R)-10,13,16-trimethyl-3,11-dioxo-17-(2,2,3,3,3-pentadeuteriopropanoyloxy)-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]ethyl] 2,2,3,3,3-pentadeuteriopropanoate
Molecular Weight (g/mol) 494.643
Formula Weight 494.309
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.