missing translation for 'onlineSavingsMsg'
Learn More

Meprednisone Acetate, TRC

Product Code. 30481650
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481650 10 mg 10mg
30481747 25 mg 25mg
30475112 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481650 Supplier Toronto Research Chemicals Supplier No. TRCM22504510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1106-03-2
Molecular Formula C24H30O6
Molecular Weight (g/mol) 414.49
Synonym 16-Meprednisone Acetate, Meprednisone 21-Acetate, (16β)-21-(Acetyloxy)-17-hydroxy-16-methyl-pregna-1,4-diene-3,11,20-trione, 17,21-Dihydroxy-16β-methyl-pregna-1,4-diene-3,11,20-trione 21-Acetate
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@@]4([C@H]3C(=O)C[C@@]2([C@]1(C(=O)COC(=O)C)O)C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H30O6/c1-13-9-18-17-6-5-15-10-16(26)7-8-22(15,3)21(17)19(27)11-23(18,4)24(13,29)20(28)12-30-14(2)25/h7-8,10,13,17-18,21,29H,5-6,9,11-12H2,1-4H3/t13-,17-,18-,21+,22-,23-,24-/m0/s1
Chemical Name or Material Meprednisone Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.