missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Merbromin

Product Code. 16402299
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402299 Supplier MedChemExpress Supplier No. HYB0961
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Merbromin acts as a topical antiseptic for minor cuts and scrapes and as a biological dye. Merbromin is a potent inhibitor against Zika virus (ZIKV) replication. Merbromin shows anti-ZIKV potency through ZIKVpro inhibition.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
Synonym Mercury dibromofluorescein disodium salt ∣ ZP1
CAS 129-16-8
Molecular Weight (g/mol) 752.67 g/mol
Solubility DMSO : 6.67 mg/mL (8.86 mM; Need ultrasonic)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.