missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic Acid, 98%

Product Code. 10716341 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10716341

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68-11-1 | C2H4O2S | 92.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-11-1
Molecular Formula C2H4O2S
Molecular Weight (g/mol) 92.11
MDL Number MFCD00004876
InChI Key CWERGRDVMFNCDR-UHFFFAOYSA-N
Synonym mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
PubChem CID 1133
ChEBI CHEBI:30065
IUPAC Name 2-sulfanylacetic acid
SMILES C(C(=O)O)S

Specifications

Melting Point -16.0°C
Color Colorless
Density 1.3250g/mL
Boiling Point 220.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CO2H
Refractive Index 1.5030 to 1.5050
Quantity 1 L
Fieser 01,1153; 07,366
Merck Index 15, 9489
Specific Gravity 1.325
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chloroform and benzene
Viscosity 6.55 Pas (20°C)
Formula Weight 92.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Mercaptoacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Mercaptoacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt