missing translation for 'onlineSavingsMsg'
Learn More

β-Mercaptoethylamine Hydrochloride, Fisher BioReagents

Product Code. 10174904
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10174904 5 g Amber Glass 5g
10666765 25 g Amber Glass 25g
10586764 100 g Amber Glass 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10174904 Supplier Fisher BioReagents Supplier No. BP26645
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
β-Mercaptoethylamine HCl is used as an antioxidant in several biochemical procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
IUPAC Name 2-aminoethanethiol;hydrochloride
SMILES [H+].[Cl-].NCCS

Specifikationer

Melting Point 66°C
Color White
pH 3.3 to 5.0
Assay Percent Range 98%
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 2776
Formula Weight 113.61
Percent Purity ∼98%
Physical Form Solid
Chemical Name or Material β-Mercaptoethylamine Hydrochloride
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 2-Aminoethanethiol hydrochloride
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.