missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Bromide Paper, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11734049 Shop All Reagecon Products
Click to view available options
:
Mercuric Bromide Paper
This item is not returnable. View return policy

Product Code. 11734049

Brand: Reagecon 1052101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to European Pharmacopoeia Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Mercuric Bromide Paper, EP Grade, Reagecon™ is a ready-to-use test sheet that has absorbed a 50g/L solution of mercuric bromide in ethanol for use in arsine/arsenic detection.

Mercuric Bromide Paper detects arsine directly at the boundary layer between water and air. White filter paper weighing 80g/m2 are placed in a 50g/L solution of mercuric bromide in ethanol, drained of excess liquid and dried. The paper turns brown when exposed to As.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions

Specifications

Certifications/Compliance Certificate of Analysis provided, According to Eu. Ph. Chapter 4(4.1.1); Prepared in ISO 9001 facility
Color White
Composition 50g/l solution of mercuric bromide in ethanol
Description Mercury dibromide (HgBr2)
CAS: 7789-47-1
Mol weight: 360.4g/mol
Format Filter paper (weighing 80g/m2)
For Use With (Application) arsine/arsenic detection
Quantity 50 Pack
Size 1.5 cm
Storage Requirements Store away from light in sealed container.
Type Mercuric Bromide Paper
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • MERCURIC BROMIDE PAPER
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, mercury dibromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.